missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylsulfonyl-4,6-dimethoxypyrimidine, 97%, ACROS Organics™

Chemical Identifiers

CAS 113583-35-0
Molecular Formula C7H10N2O4S
Molecular Weight (g/mol) 218.23
MDL Number MFCD00672151
InChI Key ITDVJJVNAASTRS-UHFFFAOYSA-N
Synonym 2-methanesulfonyl-4,6-dimethoxy-pyrimidine, 2-methanesulfonyl-4,6-dimethoxypyrimidine, 2-methylsulfony-4,6-dimethoxypyridine, 2-methylsulfonyl-4,6-dimethoxypyrimidine, 4,6-dimethoxy-2-methylsulfonyl pyrimidine, 4,6-dimethoxy-2-methylsulfonyl-pyrimidine, 4,6-dimethoxy-2-methylsulphonyl pyrimidine, maybridge1_008822, pubchem23351, pyrimidine, 4,6-dimethoxy-2-methylsulfonyl
PubChem CID 838363
IUPAC Name 2-methanesulfonyl-4,6-dimethoxypyrimidine
SMILES COC1=CC(OC)=NC(=N1)S(C)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC357280050
SDS View Documents
Thermo Fisher Scientific
AC357280050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

113583-35-0
218.23
ITDVJJVNAASTRS-UHFFFAOYSA-N
838363
COC1=CC(OC)=NC(=N1)S(C)(=O)=O
C7H10N2O4S
MFCD00672151
2-methanesulfonyl-4,6-dimethoxy-pyrimidine, 2-methanesulfonyl-4,6-dimethoxypyrimidine, 2-methylsulfony-4,6-dimethoxypyridine, 2-methylsulfonyl-4,6-dimethoxypyrimidine, 4,6-dimethoxy-2-methylsulfonyl pyrimidine, 4,6-dimethoxy-2-methylsulfonyl-pyrimidine, 4,6-dimethoxy-2-methylsulphonyl pyrimidine, maybridge1_008822, pubchem23351, pyrimidine, 4,6-dimethoxy-2-methylsulfonyl
2-methanesulfonyl-4,6-dimethoxypyrimidine

Specifications

Authentic
White
5g
96.0
97%
MFCD00672151
ITDVJJVNAASTRS-UHFFFAOYSA-N
2-methanesulfonyl-4,6-dimethoxypyrimidine
838363
Powder
2-Methylsulfonyl-4, 6-dimethoxypyrimidine
Glass Bottle
127°C to 131°C
113583-35-0
100.0
C7H10N2O4S
2-methanesulfonyl-4,6-dimethoxy-pyrimidine, 2-methanesulfonyl-4,6-dimethoxypyrimidine, 2-methylsulfony-4,6-dimethoxypyridine, 2-methylsulfonyl-4,6-dimethoxypyrimidine, 4,6-dimethoxy-2-methylsulfonyl pyrimidine, 4,6-dimethoxy-2-methylsulfonyl-pyrimidine, 4,6-dimethoxy-2-methylsulphonyl pyrimidine, maybridge1_008822, pubchem23351, pyrimidine, 4,6-dimethoxy-2-methylsulfonyl
COC1=CC(OC)=NC(=N1)S(C)(=O)=O
218.23
218.15
≥96.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications