Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Methylsulfinyl)benzoic acid, 97%

$116.92 - $346.55

Chemical Identifiers

Chemical Name or Material 2-(Methylsulfinyl)benzoic acid
CAS 19093-34-6
Molecular Formula C8H8O3S
Formula Weight 184.21
MDL Number MFCD01936003
Synonym 2-(Methanesulfinyl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2714403

  SDS
alfa aesar™
H2714403
1g Each for $116.92

AAH2714406

  SDS
alfa aesar™
H2714406
5g Each for $346.55
Description & Specifications

Specifications

Formula Weight 184.21
Chemical Name or Material 2-(Methylsulfinyl)benzoic acid
Assay 97%
Molecular Formula C8H8O3S
MDL Number MFCD01936003
CAS 19093-34-6
Synonym 2-(Methanesulfinyl)benzoic acid
Health Hazard 1 Warning
Melting Point 150° to 160°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.