Learn More
2-Methylsulfanylpyrimidine-4-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 432370010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Methylsulfanylpyrimidine-4-carboxylic acid | |
96.0 | |
Authentic | |
Glass bottle | |
MFCD09834808 | |
2-methylthio pyrimidine-4-carboxylic acid, 2-methylsulfanyl-pyrimidine-4-carboxylic acid, 2-methylthiopyrimidine-4-carboxylic acid, 2-thiomethylpyrimidine-4-carboxylic acid, 2-methylsulfanyl pyrimidine-4-carboxylic acid, 2-methylsulfanyl-pyrimidine-4-carboxylicacid, 2-methylthio-4-pyrimidinecarboxylic acid, 2-thioformylpyrimidine-4-carboxylic acid, 2-methylthio-4-pyrimidinecarboxylicacid, 2-mthylsulfanyl-pyrimidine-4-carboxylic acid | |
CSC1=NC=CC(=N1)C(O)=O | |
170.19 | |
170.19 |
1126-44-9 | |
100.0 | |
96% min. (HPLC) | |
C6H6N2O2S | |
1g | |
IAGNLKODEFUQDV-UHFFFAOYSA-N | |
2-methylsulfanylpyrimidine-4-carboxylic acid | |
295775 | |
97% |
Chemical Identifiers
1126-44-9 | |
170.19 | |
IAGNLKODEFUQDV-UHFFFAOYSA-N | |
295775 | |
CSC1=NC=CC(=N1)C(O)=O |
C6H6N2O2S | |
MFCD09834808 | |
2-methylthio pyrimidine-4-carboxylic acid, 2-methylsulfanyl-pyrimidine-4-carboxylic acid, 2-methylthiopyrimidine-4-carboxylic acid, 2-thiomethylpyrimidine-4-carboxylic acid, 2-methylsulfanyl pyrimidine-4-carboxylic acid, 2-methylsulfanyl-pyrimidine-4-carboxylicacid, 2-methylthio-4-pyrimidinecarboxylic acid, 2-thioformylpyrimidine-4-carboxylic acid, 2-methylthio-4-pyrimidinecarboxylicacid, 2-mthylsulfanyl-pyrimidine-4-carboxylic acid | |
2-methylsulfanylpyrimidine-4-carboxylic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only