Learn More
2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1267614
Description
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.
Solubility
Slightly miscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents and strong acids.
Specifications
2-Methylstyrene | |
611-15-4 | |
0.916 | |
58°C (136°F) | |
1.545 | |
25 g | |
1901764 | |
Slightly miscible with water. | |
CC1=CC=CC=C1C=C | |
118.179 | |
118.18 |
stabilized with 0.1% 4-tert-butylcatechol | |
-69°C | |
170°C to 172°C | |
C9H10 | |
MFCD00014936 | |
UN2618 | |
2-methylstyrene, 2-vinyltoluene, o-vinyltoluene, o-methylstyrene, styrene, o-methyl, 1-methyl-2-vinylbenzene, benzene, 1-ethenyl-2-methyl, ortho-methylstyrene, unii-556uo5cl44, 1-ethenyl-2-methyl-benzene | |
NVZWEEGUWXZOKI-UHFFFAOYSA-N | |
1-ethenyl-2-methylbenzene | |
11904 | |
98% |
Chemical Identifiers
611-15-4 | |
118.179 | |
NVZWEEGUWXZOKI-UHFFFAOYSA-N | |
11904 | |
CC1=CC=CC=C1C=C |
C9H10 | |
MFCD00014936 | |
2-methylstyrene, 2-vinyltoluene, o-vinyltoluene, o-methylstyrene, styrene, o-methyl, 1-methyl-2-vinylbenzene, benzene, 1-ethenyl-2-methyl, ortho-methylstyrene, unii-556uo5cl44, 1-ethenyl-2-methyl-benzene | |
1-ethenyl-2-methylbenzene |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: VINYLTOLUENES, STABILIZED
EINECSNumber : 210-256-7
RTECSNumber : WL5075900
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only