missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylresorcinol, TCI America™

Manufacturer:  TCI America M042525G

 View more versions of this product

Catalog No. M042525G



Specifications

Specifications

2-Methylresorcinol
118°C
608-25-3
MFCD00002271
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
CC1=C(C=CC=C1O)O
124.139
124.14
≥98.0% (GC)
Undesignated
25g
C7H8O2
2811
ZTMADXFOCUXMJE-UHFFFAOYSA-N
2-methylbenzene-1,3-diol
11843
Crystal-Powder at 20°C

Chemical Identifiers

608-25-3
124.139
ZTMADXFOCUXMJE-UHFFFAOYSA-N
11843
CC1=C(C=CC=C1O)O
C7H8O2
MFCD00002271
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
2-methylbenzene-1,3-diol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2766&(4)-0057

RTECSNumber : VH2009500

TSCA : Yes

Documents