missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylresorcinol 98.0+%, TCI America™

$47.73 - $353.86

Chemical Identifiers

CAS 608-25-3
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002271
InChI Key ZTMADXFOCUXMJE-UHFFFAOYSA-N
Synonym 2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
PubChem CID 11843
IUPAC Name 2-methylbenzene-1,3-diol
SMILES CC1=C(C=CC=C1O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M042525G View Documents Tci America
M042525G
25g Each for $47.73
 
M0425100G View Documents Tci America
M0425100G
100g Each for $121.80
 
M0425500G View Documents Tci America
M0425500G
500g Each for $353.86
 
Specifications

Chemical Identifiers

608-25-3
124.139
ZTMADXFOCUXMJE-UHFFFAOYSA-N
11843
CC1=C(C=CC=C1O)O
C7H8O2
MFCD00002271
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
2-methylbenzene-1,3-diol

Specifications

Undesignated
25g
C7H8O2
2811
ZTMADXFOCUXMJE-UHFFFAOYSA-N
2-methylbenzene-1,3-diol
11843
Crystal-Powder at 20°C
2-Methylresorcinol
118°C
608-25-3
MFCD00002271
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol
CC1=C(C=CC=C1O)O
124.139
124.14
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2766&(4)-0057

RTECSNumber : VH2009500

TSCA : Yes