missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylresorcinol 98.0+%, TCI America™
Supplier: TCI America M0425100G
Specifications
2-Methylresorcinol | |
118°C | |
MFCD00002271 | |
2811 | |
ZTMADXFOCUXMJE-UHFFFAOYSA-N | |
2-methylbenzene-1,3-diol | |
11843 | |
≥98.0% (GC) |
608-25-3 | |
C7H8O2 | |
100 g | |
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol | |
CC1=C(C=CC=C1O)O | |
124.139 | |
124.14 | |
Crystalline Powder |
Chemical Identifiers
608-25-3 | |
124.139 | |
ZTMADXFOCUXMJE-UHFFFAOYSA-N | |
11843 | |
CC1=C(C=CC=C1O)O |
C7H8O2 | |
MFCD00002271 | |
2-methylresorcinol, 2,6-dihydroxytoluene, 1,3-benzenediol, 2-methyl, 2-methylresorcin, resorcinol, 2-methyl, toluene-2,6-diol, 2-methyl-1,3-benzenediol, 1,3-dihydroxy-2-methylbenzene, 1,3-benzenediol, methyl, 2-methyl resorcinol | |
2-methylbenzene-1,3-diol |
Safety and Handling
EINECSNumber : (3)-2766&(4)-0057
RTECSNumber : VH2009500
TSCA : Yes