missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylquinoxaline 98.0+%, TCI America™
$64.70 - $194.10
Chemical Identifiers
CAS | 7251-61-8 |
---|---|
Molecular Formula | C9H8N2 |
Molecular Weight (g/mol) | 144.177 |
MDL Number | MFCD00006727 |
InChI Key | ALHUXMDEZNLFTA-UHFFFAOYSA-N |
Synonym | quinoxaline, 2-methyl, 2-methyl quinoxaline, unii-03vu31mv6j, ccris 2948, methyl-quinoxaline, 2-methyl-quinoxaline, acmc-1biyr, 2-methylbenzopyrazine, 2-methyl-1,4-naphthyridine, 5-23-07-00219 beilstein handbook reference |
PubChem CID | 23686 |
IUPAC Name | 2-methylquinoxaline |
SMILES | CC1=NC2=CC=CC=C2N=C1 |
Chemical Identifiers
7251-61-8 | |
144.177 | |
ALHUXMDEZNLFTA-UHFFFAOYSA-N | |
23686 | |
CC1=NC2=CC=CC=C2N=C1 |
C9H8N2 | |
MFCD00006727 | |
quinoxaline, 2-methyl, 2-methyl quinoxaline, unii-03vu31mv6j, ccris 2948, methyl-quinoxaline, 2-methyl-quinoxaline, acmc-1biyr, 2-methylbenzopyrazine, 2-methyl-1,4-naphthyridine, 5-23-07-00219 beilstein handbook reference | |
2-methylquinoxaline |
Specifications
7251-61-8 | |
C9H8N2 | |
5 g | |
ALHUXMDEZNLFTA-UHFFFAOYSA-N | |
2-methylquinoxaline | |
23686 | |
≥98.0% (GC) | |
2-Methylquinoxaline |
245°C | |
MFCD00006727 | |
quinoxaline, 2-methyl, 2-methyl quinoxaline, unii-03vu31mv6j, ccris 2948, methyl-quinoxaline, 2-methyl-quinoxaline, acmc-1biyr, 2-methylbenzopyrazine, 2-methyl-1,4-naphthyridine, 5-23-07-00219 beilstein handbook reference | |
CC1=NC2=CC=CC=C2N=C1 | |
144.177 | |
144.18 | |
Liquid |
Safety and Handling
RTECSNumber : VD3450000
TSCA : No