Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylquinoxaline, 97%

$47.77 - $610.24

Chemical Identifiers

Chemical Name or Material 2-Methylquinoxaline
CAS 7251-61-8
Molecular Formula C9H8N2
Formula Weight 144.18
MDL Number MFCD00006727
Beilstein 113307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1589509

  SDS
alfa aesar™
A1589509
10g Each for $47.77

AAA1589518

  SDS
alfa aesar™
A1589518
50g Each for $168.76

AAA1589530

  SDS
alfa aesar™
A1589530
250g Each for $610.24
Description & Specifications

Specifications

Formula Weight 144.18
Chemical Name or Material 2-Methylquinoxaline
Health Hazard 1 Warning
Molecular Formula C9H8N2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 7251-61-8
Flash Point 107°C (224°F)
Assay 97%
Beilstein 113307
Boiling Point 245° to 247°C
RTECS Number VD3450000
Quantity 10g
MDL Number MFCD00006727
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.