missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylquinoline 98.0+%, TCI America™
Supplier: TCI America Q0005100G
Specifications
2-Methylquinoline | |
-2°C | |
247°C | |
MFCD00006756 | |
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
CC1=CC=C2C=CC=CC2=N1 | |
143.19 | |
143.19 | |
Liquid |
91-63-4 | |
Yellow | |
C10H9N | |
100 g | |
SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
2-methylquinoline | |
7060 | |
≥98.0% (GC,T) |
Chemical Identifiers
91-63-4 | |
143.19 | |
SMUQFGGVLNAIOZ-UHFFFAOYSA-N | |
7060 | |
CC1=CC=C2C=CC=CC2=N1 |
C10H9N | |
MFCD00006756 | |
quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech | |
2-methylquinoline |
Safety and Handling
EINECSNumber : (5)-0795&(5)-0796
RTECSNumber : UZ9625000
TSCA : Yes