missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylquinoline N-Oxide, TCI America™

$80.97 - $241.39

Chemical Identifiers

CAS 1076-28-4
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
InChI Key FZJUONUBFWNHNU-UHFFFAOYSA-N
PubChem CID 97325
IUPAC Name 2-methyl-1-oxidoquinolin-1-ium
SMILES CC1=[N+](C2=CC=CC=C2C=C1)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M30005G View Documents TCI America
M30005G
5g Each for $241.39
 
M30001G View Documents TCI America
M30001G
1g Each for $80.97
 
Specifications

Chemical Identifiers

1076-28-4
159.188
97325
CC1=[N+](C2=CC=CC=C2C=C1)[O-]
C10H9NO
FZJUONUBFWNHNU-UHFFFAOYSA-N
2-methyl-1-oxidoquinolin-1-ium

Specifications

White-Yellow
5g
C10H9NO
CC1=[N+](C2=CC=CC=C2C=C1)[O-]
159.188
159.19
≥98.0% (GC)
67°C
1076-28-4
FZJUONUBFWNHNU-UHFFFAOYSA-N
2-methyl-1-oxidoquinolin-1-ium
97325
Crystal-Powder at 20°C
2-Methylquinoline N-Oxide
Safety and Handling

Safety and Handling

TSCA : No