missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylquinoline-7-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5498403

 View more versions of this product

Catalog No. AAH5498403



Specifications

Specifications

2-Methylquinoline-7-boronic acid pinacol ester
1g
C16H20BNO2
2-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C=CC(=N3)C
269.151
269.15
100°C to 102°C
939052-79-6
MFCD11846348
KOPSOJMEXFLBRP-UHFFFAOYSA-N
2-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
68111603
96%

Chemical Identifiers

939052-79-6
269.151
KOPSOJMEXFLBRP-UHFFFAOYSA-N
68111603
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C=CC(=N3)C
C16H20BNO2
MFCD11846348
2-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline
2-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents