Learn More
2-Methylquinoline-6-carboxylic acid, 97%, Thermo Scientific Chemicals
$29.95 - $330.75
Chemical Identifiers
CAS | 635-80-3 |
---|---|
Molecular Formula | C11H9NO2 |
Molecular Weight (g/mol) | 187.198 |
MDL Number | MFCD00014566 |
InChI Key | IZONZQFTYGVOOO-UHFFFAOYSA-N |
Synonym | 2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid |
PubChem CID | 604483 |
IUPAC Name | 2-methylquinoline-6-carboxylic acid |
SMILES | CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6379003
|
Thermo Scientific Chemicals
H6379003 |
1 g |
Each for $29.95
|
|
|||||
AAH6379006
|
Thermo Scientific Chemicals
H6379006 |
5 g |
Each for $94.67
|
|
|||||
AAH6379014
|
Thermo Scientific Chemicals
H6379014 |
25 g |
Each for $330.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
635-80-3 | |
187.198 | |
IZONZQFTYGVOOO-UHFFFAOYSA-N | |
604483 | |
CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O |
C11H9NO2 | |
MFCD00014566 | |
2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid | |
2-methylquinoline-6-carboxylic acid |
Specifications
635-80-3 | |
MFCD00014566 | |
2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid | |
CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O | |
187.198 | |
187.19 | |
Solid |
C11H9NO2 | |
1 g | |
IZONZQFTYGVOOO-UHFFFAOYSA-N | |
2-methylquinoline-6-carboxylic acid | |
604483 | |
97% | |
2-Methylquinoline-6-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only