missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylquinoline-6-carboxaldehyde, 95%

Manufacturer:  Alfa Aesar™ H6365106

 View more versions of this product

Catalog No. AAH6365106


 

Specifications

Specifications

2-Methylquinoline-6-carboxaldehyde
108166-03-6
C11H9NO
UN2811
Yellow
CC1=NC2=C(C=C1)C=C(C=C2)C=O
171.199
171.2
5g
95%
MFCD08060502
96°C to 99°C
WAPQHBBEWPVBTO-UHFFFAOYSA-N
2-methylquinoline-6-carbaldehyde
7537486
Solid

Chemical Identifiers

108166-03-6
171.199
WAPQHBBEWPVBTO-UHFFFAOYSA-N
C11H9NO
MFCD08060502
7537486
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents