missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylpyrrolidine Hydrochloride, TCI America™

$172.64 - $528.63

Chemical Identifiers

CAS 54677-53-1
Molecular Formula C5H12ClN
Molecular Weight (g/mol) 121.608
MDL Number MFCD08274961
InChI Key JNEIFWYJFOEKIM-UHFFFAOYSA-N
Synonym 2-methylpyrrolidine hydrochloride, 2-methylpyrrolidine hcl, 2-methyl-pyrrolidine hydrochloride, pubchem11112, pubchem11115, acmc-1c01e, 2-methylpyrrolidinehydrochloride, 2-methylpyrrolidine-hydrogen chloride 1/1, pyrrolidine, 2-methyl-, hydrochloride 1:1, pyrrolidine, 2-methyl-, hydrochloride, 2r-9ci
PubChem CID 21907341
IUPAC Name 2-methylpyrrolidine;hydrochloride
SMILES CC1CCCN1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21245G View Documents TCI America
M21245G
5g Each for $528.63
 
M21241G View Documents TCI America
M21241G
1g Each for $172.64
 
Specifications

Chemical Identifiers

54677-53-1
121.608
JNEIFWYJFOEKIM-UHFFFAOYSA-N
21907341
CC1CCCN1.Cl
C5H12ClN
MFCD08274961
2-methylpyrrolidine hydrochloride, 2-methylpyrrolidine hcl, 2-methyl-pyrrolidine hydrochloride, pubchem11112, pubchem11115, acmc-1c01e, 2-methylpyrrolidinehydrochloride, 2-methylpyrrolidine-hydrogen chloride 1/1, pyrrolidine, 2-methyl-, hydrochloride 1:1, pyrrolidine, 2-methyl-, hydrochloride, 2r-9ci
2-methylpyrrolidine;hydrochloride

Specifications

Undesignated
54677-53-1
MFCD08274961
JNEIFWYJFOEKIM-UHFFFAOYSA-N
2-methylpyrrolidine;hydrochloride
21907341
Crystal-Powder at 20°C
2-Methylpyrrolidine Hydrochloride
5g
C5H12ClN
2-methylpyrrolidine hydrochloride, 2-methylpyrrolidine hcl, 2-methyl-pyrrolidine hydrochloride, pubchem11112, pubchem11115, acmc-1c01e, 2-methylpyrrolidinehydrochloride, 2-methylpyrrolidine-hydrogen chloride 1/1, pyrrolidine, 2-methyl-, hydrochloride 1:1, pyrrolidine, 2-methyl-, hydrochloride, 2r-9ci
CC1CCCN1.Cl
121.608
121.61
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No