Please login to your online account to display your discounted pricing

2-Methylpyrrolidine, 98%, ACROS Organics™


Quantity 2g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC365390020

 View more versions of this product

Catalog No. AC365390020

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 85.15
Physical Form Liquid
Quantity 2g
Color Clear colorless to light yellow
Density 0.88g/mL
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2-Methylpyrrolidine
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
MDL Number MFCD00014491
Molecular Formula C5H11N
CAS Min % 97.5
Infrared Spectrum Authentic
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
CAS 765-38-8
Name Note 98%
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material 2-Methylpyrrolidine
Name Note 98%
CAS 765-38-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H11N
Formula Weight 85.15
MDL Number MFCD00014491
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents