missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylpyridine-5-boronic acid, 97%

$57.97 - $635.35

Chemical Identifiers

CAS 659742-21-9
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD06801736
InChI Key MZUSCPDSQJSBSY-UHFFFAOYSA-N
Synonym 6-methylpyridine-3-boronic acid, 2-methylpyridine-5-boronic acid, 6-methylpyridin-3-yl boronic acid, 2-methyl-5-pyridinylboronic acid, 2-picoline-5-boronic acid, 2-methyl-5-boronopyridine, boronic acid, b-6-methyl-3-pyridinyl, 6-methylpyridin-3-yl-3-boronic acid, 6-methyl-3-pyridyl boronic acid, 2-methylpyridin-5-ylboronic acid
PubChem CID 2763081
IUPAC Name (6-methylpyridin-3-yl)boronic acid
SMILES B(C1=CN=C(C=C1)C)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66327MD View Documents Alfa Aesar™
H66327MD
250mg Each for $57.97
 
AAH6632703 View Documents Alfa Aesar™
H6632703
1g Each for $166.55
 
AAH6632706 View Documents Alfa Aesar™
H6632706
5g Each for $635.35
 
Specifications

Chemical Identifiers

659742-21-9
136.945
MZUSCPDSQJSBSY-UHFFFAOYSA-N
2763081
B(C1=CN=C(C=C1)C)(O)O
C6H8BNO2
MFCD06801736
6-methylpyridine-3-boronic acid, 2-methylpyridine-5-boronic acid, 6-methylpyridin-3-yl boronic acid, 2-methyl-5-pyridinylboronic acid, 2-picoline-5-boronic acid, 2-methyl-5-boronopyridine, boronic acid, b-6-methyl-3-pyridinyl, 6-methylpyridin-3-yl-3-boronic acid, 6-methyl-3-pyridyl boronic acid, 2-methylpyridin-5-ylboronic acid
(6-methylpyridin-3-yl)boronic acid

Specifications

Brown to White
659742-21-9
MFCD06801736
MZUSCPDSQJSBSY-UHFFFAOYSA-N
(6-methylpyridin-3-yl)boronic acid
2763081
Powder
2-Methylpyridine-5-boronic acid
250mg
C6H8BNO2
6-methylpyridine-3-boronic acid, 2-methylpyridine-5-boronic acid, 6-methylpyridin-3-yl boronic acid, 2-methyl-5-pyridinylboronic acid, 2-picoline-5-boronic acid, 2-methyl-5-boronopyridine, boronic acid, b-6-methyl-3-pyridinyl, 6-methylpyridin-3-yl-3-boronic acid, 6-methyl-3-pyridyl boronic acid, 2-methylpyridin-5-ylboronic acid
B(C1=CN=C(C=C1)C)(O)O
136.945
136.94
97%