Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylpyridine-4-boronic acid pinacol ester, 95%, may contain up to 3% water

$406.36 - $931.27

Chemical Identifiers

Chemical Name or Material 2-Methylpyridine-4-boronic acid pinacol ester
CAS 660867-80-1
Molecular Formula C12H18BNO2
Formula Weight 219.09
MDL Number MFCD08061590
Synonym 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2745003

 
alfa aesar™
H2745003
1g Each for $931.27

AAH27450MD

 
alfa aesar™
H27450MD
250mg Each for $406.36
Description & Specifications

Specifications

Formula Weight 219.09
Chemical Name or Material 2-Methylpyridine-4-boronic acid pinacol ester
Assay 95%
Assay Percent Notes may contain up to 3% water
Molecular Formula C12H18BNO2
MDL Number MFCD08061590
CAS 660867-80-1
Synonym 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Warning
Melting Point 66° to 71°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C