missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylpyrazine, TCI America™

$38.19 - $192.50

Chemical Identifiers

CAS 109-08-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006142
InChI Key CAWHJQAVHZEVTJ-UHFFFAOYSA-N
Synonym methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
PubChem CID 7976
IUPAC Name 2-methylpyrazine
SMILES CC1=NC=CN=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M073625ML View Documents TCI America
M073625ML
25mL Each for $38.19
 
M0736250ML View Documents TCI America
M0736250ML
250mL Each for $192.50
 
Specifications

Chemical Identifiers

109-08-0
94.117
CAWHJQAVHZEVTJ-UHFFFAOYSA-N
7976
CC1=NC=CN=C1
C5H6N2
MFCD00006142
methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
2-methylpyrazine

Specifications

135°C
-29°C
109-08-0
MFCD00006142
methylpyrazine, pyrazine, methyl, pyrazine, 2-methyl, 2-methyl-1,4-diazine, 2-methyl pyrazine, 2-methyl-pyrazine, unii-rvc6500u9c, fema no. 3309, ccris 2927, wln: t6n dnj b1
CC1=NC=CN=C1
94.117
94.12
≥98.0% (GC)
Yellow
25mL
C5H6N2
1993
CAWHJQAVHZEVTJ-UHFFFAOYSA-N
2-methylpyrazine
7976
Clear Liquid at 20°C
2-Methylpyrazine
Safety and Handling

Safety and Handling

RTECSNumber : UQ3675000

TSCA : Yes