Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylpyrazine, 99+%

Chemical Identifiers

Chemical Name or Material 2-Methylpyrazine
CAS 109-08-0
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00006142
UN Number UN1993
Beilstein 105778
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1414614

 
alfa aesar™
A1414614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1414622

 
alfa aesar™
A1414622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1414636

 
alfa aesar™
A1414636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 94.12
Chemical Name or Material 2-Methylpyrazine
UN Number UN1993
Health Hazard 2 GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point −30°C
Flash Point 50°C (122°F)
Quantity 25g
Odor Unpleasant
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥99%
Beilstein 105778
Boiling Point 135° to 136°C
TSCA TSCA
Molecular Formula C5H6N2
CAS 109-08-0
RTECS Number UQ3675000
MDL Number MFCD00006142
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Health Hazard 1 Danger

Applications
2-Methylpyrazine is used as flavoring agents in bread fresh bread, butter, chocolate, chocolate cocoa, coffee, meat roasted, nut almond.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.