Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylpyrazine-5-carboxylic acid, 98%

$27.12 - $170.27

Chemical Identifiers

Chemical Name or Material 2-Methylpyrazine-5-carboxylic acid
CAS 5521-55-1
Molecular Formula C6H6N2O2
Formula Weight 138.13
MDL Number MFCD00068241
Beilstein 116005
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1401103

  SDS
alfa aesar™
A1401103
1g Each for $27.12

AAA1401106

  SDS
alfa aesar™
A1401106
5g Each for $52.88

AAA1401114

  SDS
alfa aesar™
A1401114
25g Each for $170.27
Description & Specifications

Specifications

Formula Weight 138.13
Chemical Name or Material 2-Methylpyrazine-5-carboxylic acid
Assay 98%
Molecular Formula C6H6N2O2
MDL Number MFCD00068241
CAS 5521-55-1
Beilstein 116005
Health Hazard 1 Danger
Melting Point 165° to 167°C
Quantity 1g
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.