missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylpiperidine 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 127495000

 View more versions of this product

Catalog No. AC127495000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Liquid
Clear yellow
0.8440g/mL
1.4459 to 1.4479 (20°C, 589nm)
98.5
2-Pipecoline
99%
99%
109-05-7
99.18
500mL
20, 95
100.0
Authentic
Glass bottle
2-Methylpiperidine
C6H13N
MFCD00005982

Chemical Identifiers

109-05-7
100.0
C6H13N
MFCD00005982
2-Methylpiperidine, 99%
98.5
99%
99.18
2-Pipecoline
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-642-1

RTECSNumber : TN1240000

TSCA : TSCA