Please login to your online account to display your discounted pricing

2-Methylphenylacetone, 97%, ACROS Organics™

$153.89 - $153.89

Chemical Identifiers

Chemical Name or Material 2-Methylphenylacetone
Name Note 97%
CAS 51052-00-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10 H12 O
Formula Weight 148.21
MDL Number MFCD00210465
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351120050

  SDS
thermo fisher scientific
351120050
Glass bottle 5g Each for $153.89
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 148.21
Physical Form Liquid
Quantity 5g
Color Clear light yellow
TSCA TSCA
Beilstein 07, IV, 721
CAS Max % 100.0
CAS Min % 96.0
Packaging Glass bottle
Refractive Index 1.5185 to 1.5205 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material 2-Methylphenylacetone
Name Note 97%
MDL Number MFCD00210465
Molecular Formula C10 H12 O
CAS 51052-00-7
Assay Percent Range 97%