Please login to your online account to display your discounted pricing

(2-Methylphenyl)methanesulfonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material (2-Methylphenyl)methanesulfonyl chloride
CAS 92614-55-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9ClO2S
Formula Weight 204.68
MDL Number MFCD02683108
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07798DA

  SDS
fisher scientific company
MO07798DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.68
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material (2-Methylphenyl)methanesulfonyl chloride
Molecular Formula C8H9ClO2S
MDL Number MFCD02683108
CAS 92614-55-6
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.