missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Methylphenoxy)acetic acid, 99%, ACROS Organics™

Chemical Identifiers

CAS 1878-49-5
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C9 H10 O3
Linear Formula CH3C6H4OCH2CO2H
Formula Weight 166.18
MDL Number MFCD00014354
Chemical Name or Material (2-Methylphenoxy)acetic acid, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC364250250 SDS Thermo Fisher Scientific
AC364250250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1878-49-5
100.0
C9 H10 O3
166.18
(2-Methylphenoxy)acetic acid, 99%
98.5
99%
CH3C6H4OCH2CO2H
MFCD00014354
≥98.5%

Specifications

White
25g
98.5
Authentic
Needle-Like Crystals
C9 H10 O3
166.18
(2-Methylphenoxy)acetic acid, 99%
154° to 157°C
1878-49-5
100.0
Glass bottle
99%
CH3C6H4OCH2CO2H
MFCD00014354
≥98.5%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.