missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylpentane 98.0+%, TCI America™

Manufacturer:  TCI America M038225ML

 View more versions of this product

Catalog No. M038225ML



Specifications

Specifications

2-Methylpentane
Colorless
107-83-5
MFCD00009406
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
CCCC(C)C
86.178
86.18
≥98.0% (GC)
60°C
25mL
C6H14
1208
AFABGHUZZDYHJO-UHFFFAOYSA-N
2-methylpentane
7892
Clear Liquid at 20°C

Chemical Identifiers

107-83-5
86.178
AFABGHUZZDYHJO-UHFFFAOYSA-N
7892
CCCC(C)C
C6H14
MFCD00009406
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
2-methylpentane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0006

RTECSNumber : SA2985000

TSCA : Yes

Documents