Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylnaphtho[2,3-d]oxazole, 98%

$95.74 - $350.96

Chemical Identifiers

Chemical Name or Material 2-Methylnaphtho[2,3-d]oxazole
CAS 20686-66-2
Molecular Formula C12H9NO
Formula Weight 183.21
MDL Number MFCD00624597
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2180606

 
alfa aesar™
B2180606
5g Each for $95.74

AAB2180614

 
alfa aesar™
B2180614
25g Each for $350.96
Description & Specifications

Specifications

Formula Weight 183.21
Molecular Formula C12H9NO
Chemical Name or Material 2-Methylnaphtho[2,3-d]oxazole
Assay 98%
MDL Number MFCD00624597
CAS 20686-66-2
Melting Point 85° to 88°C
Boiling Point 309° to 311°C
Quantity 5g