missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylnaphthalene 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 127175000

 View more versions of this product

Catalog No. AC127175000


 

Specifications Description & Specifications

Specifications

2-Methylnaphthalene
91-57-6
1.01g/mL
Glass bottle
≥96%
C11H10
142.2
05, 567
32° to 36°C
96%
96
Authentic
Crystalline Low Melting Solid
95 to 97%
C10H7CH3
MFCD00004118
White
500g

Chemical Identifiers

91-57-6
95 to 97%
C10H7CH3
MFCD00004118
≥96%
96
C11H10
142.2
2-Methylnaphthalene, 96%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-078-3

RTECSNumber : QJ9635000

TSCA : TSCA