Please login to your online account to display your discounted pricing

2-Methylnaphth[2,1-d]oxazole, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  414580250

 View more versions of this product

Catalog No. AC414580250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Formula Weight 183.21
Physical Form Liquid
Quantity 25g
Color Clear yellow-brown to orange-brown
Beilstein 27, IV, 1342
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Name Note 96%
Refractive Index 1.6340 to 1.6370 (20°C, 589nm)
Assay Percent Range 96%
Molecular Formula C12 H9 N O
MDL Number MFCD00022185
Chemical Name or Material 2-Methylnaphth[2, 1-d]oxazole
Infrared Spectrum Authentic
CAS 20686-65-1

Specifications

Chemical Name or Material 2-Methylnaphth[2, 1-d]oxazole
Name Note 96%
CAS 20686-65-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C12 H9 N O
Formula Weight 183.21
MDL Number MFCD00022185