Please login to your online account to display your discounted pricing

2-Methylnaphth[2,1-d]oxazole, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methylnaphth[2, 1-d]oxazole
Name Note 96%
CAS 20686-65-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C12 H9 N O
Formula Weight 183.21
MDL Number MFCD00022185
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC414580050

  SDS
thermo fisher scientific
AC414580050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC414580250

  SDS
thermo fisher scientific
414580250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 183.21
Quantity 5g
Color Clear yellow-brown to orange-brown
Beilstein 27, IV, 1342
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.6340 to 1.6370 (20°C, 589nm)
Infrared Spectrum Authentic
Assay Percent Range 96%
Molecular Formula C12 H9 N O
Chemical Name or Material 2-Methylnaphth[2, 1-d]oxazole
Name Note 96%
CAS 20686-65-1
MDL Number MFCD00022185