missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Methylmercapto)benzimidazole 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 127121000

 View more versions of this product

Catalog No. AC127121000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Fine Crystalline Powder
Light yellow to beige
100.0
98.5
2-(Methylmercapto)benzimidazole
C8H8N2S
7152-24-1
164.22
100g
202° to 205°C
Authentic
MFCD00005594
99%
2-(Methylthio)benzimidazole

Chemical Identifiers

7152-24-1
100.0
164.22
2-(Methylthio)benzimidazole
≥98.5%
98.5
C8H8N2S
MFCD00005594
2-(Methylmercapto)benzimidazole, 99%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 230-494-5

RTECSNumber : DD9480000

TSCA : TSCA