Please login to your online account to display your discounted pricing

2-(Methylmercapto)benzimidazole 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Methylmercapto)benzimidazole
Name Note 99%
CAS 7152-24-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Molecular Formula C8H8N2S
Formula Weight 164.22
MDL Number MFCD00005594
Synonym 2-(Methylthio)benzimidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC127121000

  SDS
thermo fisher scientific
127121000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 164.22
Physical Form Fine Crystalline Powder
Quantity 100g
Color Light yellow to beige
EINECS Number 230-494-5
RTECS Number DD9480000
TSCA TSCA
Melting Point 202° to 205°C
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 98.5
MDL Number MFCD00005594
Chemical Name or Material 2-(Methylmercapto)benzimidazole
Name Note 99%
Molecular Formula C8H8N2S
Synonym 2-(Methylthio)benzimidazole
CAS 7152-24-1