Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylmalonamide, 95%

$76.39 - $277.24

Chemical Identifiers

Chemical Name or Material 2-Methylmalonamide
CAS 1113-63-9
Molecular Formula C4H8N2O2
Formula Weight 116.12
MDL Number MFCD00043625
Beilstein 1756100
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0920304

  SDS
alfa aesar™
L0920304
2g Each for $76.39

AAL0920309

  SDS
alfa aesar™
L0920309
10g Each for $277.24
Description & Specifications

Specifications

Formula Weight 116.12
CAS 1113-63-9
Molecular Formula C4H8N2O2
Assay 95%
MDL Number MFCD00043625
Beilstein 1756100
Quantity 2g
Melting Point 208° to 210°C
Chemical Name or Material 2-Methylmalonamide