missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylindoline, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 275880050

 View more versions of this product

Catalog No. AC275880050



Description

Description

Specifications

Specifications

2-Methylindoline
103°C
Glass bottle
1.02
227°C (1013.0 mbar)
-51°C
99+%
99.0
99+%
MFCD00005706
.alpha.-methyldihydroindole, 1h-indole, 2,3-dihydro-2-methyl, 2,3-dihydro-2-methyl-1h-indole, 2,3-dihydro-2-methylindole, 2-methyl-2,3-dihydro-1h-indole, 2-methyl-2,3-dihydroindole, 2-methylindoline, alpha-methyldihydroindole, dihydroindole, 2-methyl, indoline, 2-methyl
QRWRJDVVXAXGBT-UHFFFAOYNA-N
23305
Liquid
1.0200g/mL
Authentic
1.5680 to 1.57
0.5% max. (K.F.)
Yellow to Brown
5mL
6872-06-6
100.0
C9H11N
20, 279
Solubility in water: negligible
133.19
133.19
99+%

Chemical Identifiers

6872-06-6
133.19
QRWRJDVVXAXGBT-UHFFFAOYNA-N
23305
C9H11N
MFCD00005706
.alpha.-methyldihydroindole, 1h-indole, 2,3-dihydro-2-methyl, 2,3-dihydro-2-methyl-1h-indole, 2,3-dihydro-2-methylindole, 2-methyl-2,3-dihydro-1h-indole, 2-methyl-2,3-dihydroindole, 2-methylindoline, alpha-methyldihydroindole, dihydroindole, 2-methyl, indoline, 2-methyl
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-971-0

RTECSNumber : NM1926350

TSCA : TSCA

SDS Documents