missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylindole-3-carboxamide, TCI America™

$247.50 - $722.66

Chemical Identifiers

CAS 67242-60-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00143101
InChI Key QKWLFJZXUBMFLU-UHFFFAOYSA-N
Synonym 3-Carbamoyl-2-methylindole
PubChem CID 12415229
IUPAC Name 2-methyl-1H-indole-3-carboxamide
SMILES CC1=C(C2=CC=CC=C2N1)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C12551G View Documents TCI America
C12551G
1g Each for $247.50
 
C12555G View Documents TCI America
C12555G
5g Each for $722.66
 
Specifications

Chemical Identifiers

67242-60-8
174.203
QKWLFJZXUBMFLU-UHFFFAOYSA-N
12415229
CC1=C(C2=CC=CC=C2N1)C(=O)N
C10H10N2O
MFCD00143101
3-Carbamoyl-2-methylindole
2-methyl-1H-indole-3-carboxamide

Specifications

White
1g
C10H10N2O
3-Carbamoyl-2-methylindole
CC1=C(C2=CC=CC=C2N1)C(=O)N
174.203
174.20
≥98.0% (T)
225°C
67242-60-8
MFCD00143101
QKWLFJZXUBMFLU-UHFFFAOYSA-N
2-methyl-1H-indole-3-carboxamide
12415229
Crystal-Powder at 20°C
2-Methylindole-3-carboxamide
Safety and Handling

Safety and Handling

TSCA : No