Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylimidazole-1-propionic acid, 97%

$366.15 - $366.15

Chemical Identifiers

Chemical Name or Material 2-Methylimidazole-1-propionic acid
CAS 24647-62-9
Molecular Formula C7H10N2O2
Formula Weight 154.17
MDL Number MFCD00234356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5022103

  SDS
alfa aesar™
H5022103
1g Each for $366.15

AAH50221MD

  SDS
alfa aesar™
H50221MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Methylimidazole-1-propionic acid
CAS 24647-62-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Molecular Formula C7H10N2O2
Formula Weight 154.17
MDL Number MFCD00234356
Quantity 1g

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water (250 mg/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.