missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazo[4,5-c]pyridine, 96%

CAS: 63604-59-1 | C7H7N3 | 133.154 g/mol

$83.14 - $413.08

Chemical Identifiers

CAS 63604-59-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD03426025
InChI Key JWQVOMJORQXNME-UHFFFAOYSA-N
Synonym 2-methyl-3h-imidazo 4,5-c pyridine, 2-methyl-1h-imidazo 4,5-c pyridine, 2-methylimidazo 4,5-c pyridine, 8-methyl-3-deazapurine, 8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, acmc-20anq0, 2-methyl-5-azabenzimidazole, 3h-imidazo 4,5-c pyridine, 2-methyl, 1h-imidazo 4,5-c pyridine, 2-methyl
PubChem CID 373077
IUPAC Name 2-methyl-3H-imidazo[4,5-c]pyridine
SMILES CC1=NC2=C(N1)C=NC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL19131MD
View Documents
Thermo Scientific Chemicals
L19131MD
250 mg N/A
Only null left
 
AAL1913103
View Documents
Thermo Scientific Chemicals
L1913103
1 g N/A
Only null left
 
AAL1913106
View Documents
Thermo Scientific Chemicals
L1913106
5 g N/A
Only null left
 
Description

Description

2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

63604-59-1
133.154
JWQVOMJORQXNME-UHFFFAOYSA-N
373077
CC1=NC2=C(N1)C=NC=C2
C7H7N3
MFCD03426025
2-methyl-3h-imidazo 4,5-c pyridine, 2-methyl-1h-imidazo 4,5-c pyridine, 2-methylimidazo 4,5-c pyridine, 8-methyl-3-deazapurine, 8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, acmc-20anq0, 2-methyl-5-azabenzimidazole, 3h-imidazo 4,5-c pyridine, 2-methyl, 1h-imidazo 4,5-c pyridine, 2-methyl
2-methyl-3H-imidazo[4,5-c]pyridine

Specifications

63604-59-1
C7H7N3
250 mg
2-methyl-3h-imidazo 4,5-c pyridine, 2-methyl-1h-imidazo 4,5-c pyridine, 2-methylimidazo 4,5-c pyridine, 8-methyl-3-deazapurine, 8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, acmc-20anq0, 2-methyl-5-azabenzimidazole, 3h-imidazo 4,5-c pyridine, 2-methyl, 1h-imidazo 4,5-c pyridine, 2-methyl
JWQVOMJORQXNME-UHFFFAOYSA-N
2-methyl-3H-imidazo[4,5-c]pyridine
373077
99%
176°C to 178°C
MFCD03426025
3234
Insoluble in water.
CC1=NC2=C(N1)C=NC=C2
133.154
133.15
2-Methylimidazo[4,5-c]pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only