missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC21701DE

 View more versions of this product

Catalog No. CC21701DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

176.17
97%
C9H8N2O2
2-Methylimidazo[1, 2-a]pyridine-3-carboxylic acid
96.5
5g
100.0
97%
21801-79-6
MFCD00244615

Chemical Identifiers

21801-79-6
100.0
C9H8N2O2
MFCD00244615
97%
96.5
97%
176.17
2-Methylimidazo[1, 2-a]pyridine-3-carboxylic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.