Learn More
2-Methylhydroquinone, 99%, Thermo Scientific Chemicals
$115.63 - $347.79
Chemical Identifiers
CAS | 95-71-6 |
---|---|
Molecular Formula | C7H8O2 |
Molecular Weight (g/mol) | 124.14 |
MDL Number | MFCD00002345 |
InChI Key | CNHDIAIOKMXOLK-UHFFFAOYSA-N |
Synonym | methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol |
PubChem CID | 7253 |
IUPAC Name | 2-methylbenzene-1,4-diol |
SMILES | CC1=CC(O)=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2124130
|
Thermo Scientific Chemicals
B2124130 |
250 g |
Each for $115.63
|
|
|||||
AAB212410B
|
Thermo Scientific Chemicals
B212410B |
1000 g |
Each for $347.79
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-71-6 | |
124.14 | |
CNHDIAIOKMXOLK-UHFFFAOYSA-N | |
7253 | |
CC1=CC(O)=CC=C1O |
C7H8O2 | |
MFCD00002345 | |
methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol | |
2-methylbenzene-1,4-diol |
Specifications
95-71-6 | |
284°C to 286°C | |
C7H8O2 | |
250 g | |
methylhydroquinone, 2-methylhydroquinone, 2,5-dihydroxytoluene, p-toluhydroquinone, toluhydroquinone, toluquinol, p-toluquinol, tolylhydroquinone, p-toluhydroquinol, 2,5-toluenediol | |
CC1=CC(O)=CC=C1O | |
124.14 | |
124.14 | |
2-Methylhydroquinone |
127°C to 130°C | |
172°C (341°F) | |
MFCD00002345 | |
2041489 | |
CNHDIAIOKMXOLK-UHFFFAOYSA-N | |
2-methylbenzene-1,4-diol | |
7253 | |
99% |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
H302-H317-H318-H341-H351
EINECSNumber : 202-443-7
RTECSNumber : MX6700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only