Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methylhexanoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Methylhexanoyl chloride
CAS 41693-47-4
Molecular Formula C7H13ClO
Formula Weight 148.63
MDL Number MFCD01318644
UN Number UN2920
Synonym 2-Methylcaproyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3282903

  SDS
alfa aesar™
H3282903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3282909

  SDS
alfa aesar™
H3282909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.63
Assay 97%
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
UN Number UN2920
Molecular Formula C7H13ClO
CAS 41693-47-4
Boiling Point 163°C
Chemical Name or Material 2-Methylhexanoyl chloride
Quantity 1g
Flash Point 55°C (131°F)
MDL Number MFCD01318644
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Health Hazard 1 Danger
Synonym 2-Methylcaproyl chloride
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.