Please login to your online account to display your discounted pricing

2-Methylhexanoic acid, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  269420250

 View more versions of this product

Catalog No. AC269420250

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 0.9160g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4180 to 1.4220 (20°C, 589nm)
Color Clear colorless to slightly yellow
Quantity 25g
Chemical Name or Material 2-Methylhexanoic acid
Name Note 98%
CAS 4536-23-6
CAS Min % 97.5
CAS Max % 100.0
Physical Form Clear colorless to slightly yellow liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C7H14O2
Linear Formula CH3(CH2)3CH(CH3)CO2H
Formula Weight 130.19g/mol
MDL Number MFCD00002674
Synonym 2-Methylcaproic acid
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number MO8400600
EINECS Number 224-883-9
TSCA TSCA

Specifications

Chemical Name or Material 2-Methylhexanoic acid
Name Note 98%
CAS 4536-23-6
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C7H14O2
Linear Formula CH3(CH2)3CH(CH3)CO2H
Formula Weight 130.19g/mol
MDL Number MFCD00002674
Synonym 2-Methylcaproic acid
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents