missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylhexanoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 269420050

 View more versions of this product

Catalog No. AC269420050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear colorless to slightly yellow
100.0
Glass bottle
1.4180 to 1.4220 (20°C, 589nm)
98%
C7H14O2
2-Methylhexanoic acid
MFCD00002674
130.19
5g
0.9160g/mL
97.5
Authentic
2-Methylcaproic acid
98%
4536-23-6
CH3(CH2)3CH(CH3)CO2H

Chemical Identifiers

4536-23-6
100.0
C7H14O2
130.19
2-Methylcaproic acid
≥97.5%
97.5
98%
CH3(CH2)3CH(CH3)CO2H
MFCD00002674
2-Methylhexanoic acid, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-883-9

RTECSNumber : MO8400600

TSCA : TSCA