missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylhexanoic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 4536-23-6
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H14O2
Linear Formula CH3(CH2)3CH(CH3)CO2H
Formula Weight 130.19
MDL Number MFCD00002674
Synonym 2-Methylcaproic acid
Chemical Name or Material 2-Methylhexanoic acid, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC269420050 SDS Thermo Fisher Scientific
269420050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

4536-23-6
100.0
C7H14O2
130.19
2-Methylcaproic acid
≥97.5%
97.5
98%
CH3(CH2)3CH(CH3)CO2H
MFCD00002674
2-Methylhexanoic acid, 98%

Specifications

130.19
5g
0.9160g/mL
97.5
Authentic
2-Methylcaproic acid
C7H14O2
CH3(CH2)3CH(CH3)CO2H
2-Methylhexanoic acid, 98%
Liquid
Clear colorless to slightly yellow
100.0
Glass bottle
1.4180 to 1.4220 (20°C, 589nm)
98%
4536-23-6
MFCD00002674
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 224-883-9

RTECSNumber : MO8400600

TSCA : TSCA