Learn More
2-Methylhexanoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1129006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methylhexanoic acid | |
0.916 | |
105°C (221°F) | |
C7H13O2 | |
MFCD00002674 | |
UN3265 | |
2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl | |
CCCC[C@@H](C)C([O-])=O | |
129.18 | |
130.19 |
4536-23-6 | |
209°C to 210°C | |
Rancid | |
1.4205 | |
5 g | |
1721227 | |
CVKMFSAVYPAZTQ-ZCFIWIBFSA-M | |
2-methylhexanoic acid | |
20653 | |
98% |
Chemical Identifiers
4536-23-6 | |
129.18 | |
CVKMFSAVYPAZTQ-ZCFIWIBFSA-M | |
20653 |
C7H13O2 | |
MFCD00002674 | |
2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl | |
CCCC[C@@H](C)C([O-])=O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 224-883-9
RTECSNumber : MO8400600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only