missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methylhexane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 126910050

 View more versions of this product

Catalog No. AC126910050



Description

Description

Specifications

Specifications

2-Methylhexane
−3°C
Glass bottle
0.67
Colorless
5mL
98.5
98.5% min. (GC)
CH3(CH2)3CH(CH3)2
01, 156
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,miscible with ether,acetone
CCCCC(C)C
100.2
100.2
99%
0.6700g/mL
Authentic
1.3835 to 1.3855
90.0°C
-118.0°C
591-76-4
100.0
C7H16
MFCD00009487
2-methyl hexane, 2-methylhexane, 5-methylhexan-2-yl, 5-methylhexan-3-yl, acmc-1b07e, hexane, 2-methyl, isoheptane, isoheptanes, pentane, dimethyl, unii-yi4679p2sp
GXDHCNNESPLIKD-UHFFFAOYSA-N
2-methylhexane
11582
Liquid

Chemical Identifiers

591-76-4
100.2
GXDHCNNESPLIKD-UHFFFAOYSA-N
11582
CCCCC(C)C
C7H16
MFCD00009487
2-methyl hexane, 2-methylhexane, 5-methylhexan-2-yl, 5-methylhexan-3-yl, acmc-1b07e, hexane, 2-methyl, isoheptane, isoheptanes, pentane, dimethyl, unii-yi4679p2sp
2-methylhexane
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-730-6

SDS Documents