Learn More
2-Methylhexane, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 126910250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Methylhexane | |
98.5 | |
0.6700g/mL | |
−3°C | |
98.5% min. (GC) | |
C7H16 | |
CH3(CH2)3CH(CH3)2 | |
25mL | |
0.67 | |
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,miscible with ether,acetone | |
CCCCC(C)C | |
100.21 | |
100.2 |
591-76-4 | |
100.0 | |
90.0°C | |
Authentic | |
Glass bottle | |
1.3835 to 1.3855 | |
MFCD00009487 | |
01, 156 | |
hexane, 2-methyl, isoheptane, unii-yi4679p2sp, pentane, dimethyl, isoheptanes, 2-methyl hexane, 2-methylhexane, acmc-1b07e, 5-methylhexan-2-yl, 5-methylhexan-3-yl | |
GXDHCNNESPLIKD-UHFFFAOYSA-N | |
2-methylhexane | |
11582 | |
99% |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-730-6
RUO â Research Use Only