missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methylheptanoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2205022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methylheptanoic acid | |
0.906 | |
>110°C (230°F) | |
1.4255 | |
100 g | |
NKBWMBRPILTCRD-UHFFFAOYSA-N | |
2-methylheptanoic acid | |
14475 | |
98% |
1188-02-9 | |
139°C to 141°C (30 mmHg) | |
C8H16O2 | |
MFCD00142944 | |
2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s | |
CCCCCC(C)C(=O)O | |
144.214 | |
144.22 |
Chemical Identifiers
1188-02-9 | |
144.214 | |
NKBWMBRPILTCRD-UHFFFAOYSA-N | |
14475 | |
CCCCCC(C)C(=O)O |
C8H16O2 | |
MFCD00142944 | |
2-methylenanthic acid, heptanoic acid, 2-methyl, methylamylacetic acid, 2-methyl-heptanoic acid, heptanoic acid, methyl, alpha-methylheptanoic acid, 2-methyl heptanoic acid, heptane-2-carboxylic acid, fema no. 2706, heptanoic acid, 2-methyl-, 2s | |
2-methylheptanoic acid |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 214-704-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only