Please login to your online account to display your discounted pricing

2-Methylglutaric acid, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  295380050

 View more versions of this product

Catalog No. AC295380050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 146.14
Quantity 5g
Color White to almost white
TSCA TSCA
Melting Point 75° to 79°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
Chemical Name or Material 2-Methylglutaric acid
Assay Percent Range 99%
CAS 18069-17-5
Name Note 99%
MDL Number MFCD00002661
Linear Formula HO2CCH2CH2CH(CH3)CO2H
Molecular Formula C6H10O4

Specifications

Chemical Name or Material 2-Methylglutaric acid
Name Note 99%
CAS 18069-17-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H10O4
Linear Formula HO2CCH2CH2CH(CH3)CO2H
Formula Weight 146.14
MDL Number MFCD00002661