Please login to your online account to display your discounted pricing

2-Methylglutaric acid, 99%, ACROS Organics™

$31.45 - $31.45

Chemical Identifiers

Chemical Name or Material 2-Methylglutaric acid
Name Note 99%
CAS 18069-17-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H10O4
Linear Formula HO2CCH2CH2CH(CH3)CO2H
Formula Weight 146.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC295380050

  SDS
thermo fisher scientific
295380050
Glass bottle 5g Each for $31.45
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 146.14
Quantity 5g
Color White to almost white
TSCA TSCA
Melting Point 75° to 79°C
CAS Max % 100.0
Chemical Name or Material 2-Methylglutaric acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H10O4
Linear Formula HO2CCH2CH2CH(CH3)CO2H
MDL Number MFCD00002661
CAS 18069-17-5
CAS Min % 98.5