Learn More
2-Methylfuran, 99%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126830050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methylfuran | |
534-22-5,123-31-9 | |
100.0 | |
Yellow | |
63.0°C to 66.0°C | |
Authentic | |
Glass bottle | |
1.4320 to 1.4340 | |
5 mL | |
01,682; 14,209 | |
methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran | |
VQKFNUFAXTZWDK-UHFFFAOYSA-N | |
2-methylfuran | |
0.01% hydroquinone | |
4 mPa.s (25°C) | |
99% |
Stabilized | |
98.5 | |
-88.7°C | |
0.9100g/mL | |
−26°C | |
98.5% min. (GC) | |
C5H6O | |
MFCD00003248 | |
17, 36 | |
0.91 | |
Solubility in water: 0.3g/100mL(20 c). Other solubilities: miscible with most common organic solvents | |
CC1=CC=CO1 | |
82.1 | |
10797 | |
82.1 | |
Liquid |
Chemical Identifiers
534-22-5 | |
VQKFNUFAXTZWDK-UHFFFAOYSA-N | |
10797 | |
CC1=CC=CO1 |
MFCD00003248 | |
methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran | |
2-methylfuran |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-594-5
RUO – Research Use Only